missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Prolactin Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 537.00
Specifications
| Antigen | Prolactin |
|---|---|
| Clone | 1085531 |
| Dilution | Western Blot 0.5 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry, Western Blot, Simple Western |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30241035
|
R&D Systems
MAB11590-100 |
100 μg |
€ 537.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30241023
|
R&D Systems
MAB11590-SP |
25 μg |
€ 214.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple WesternSpecifications
| Prolactin | |
| Western Blot 0.5 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| PRL, prolactin | |
| E. coli- derived human Prolactin, Leu29-Cys227, Accession # P01236 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1085531 | |
| Immunohistochemistry, Western Blot, Simple Western | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01236 | |
| 5617 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title