Learn More
Human TAPBPR Antibody, Novus Biologicals™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11337-SP
 View more versions of this product
Product Code. 18795965
  • € 242.00 / 25µg
Description
Tapasin Related Protein Monoclonal antibody specifically detects Tapasin Related Protein in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tapasin Related Protein
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBPL, TAPBPR, TAPBP-RTAP binding protein related, TAPBPRtapasin-like
Chinese Hamster Ovary cell line, CHO-derived human TAPBPR, Ala19-Arg404, Accession # Q9BX59.2
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a
Flow Cytometry
1059329
Flow Cytometry 0.25 ug/10^6 cells
Q9BX59.2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
55080