missing translation for 'onlineSavingsMsg'
Learn More

Human TAPBPR Antibody, Novus Biologicals™
GREENER_CHOICE

Mouse Monoclonal Antibody

€ 242.00 - € 686.00

Specifications

Antigen Tapasin Related Protein
Clone 1059329
Dilution Flow Cytometry 0.25 ug/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18776125
GREENER_CHOICE
View Documents
R&D Systems
MAB11337-100
100 μg
€ 686.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18795965
GREENER_CHOICE
View Documents
R&D Systems
MAB11337-SP
25 μg
€ 242.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tapasin Related Protein Monoclonal antibody specifically detects Tapasin Related Protein in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

Tapasin Related Protein
Flow Cytometry 0.25 ug/10^6 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBPL, TAPBPR, TAPBP-RTAP binding protein related, TAPBPRtapasin-like
Chinese Hamster Ovary cell line, CHO-derived human TAPBPR, Ala19-Arg404, Accession # Q9BX59.2
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1059329
Flow Cytometry
Unconjugated
Mouse
Human
Q9BX59.2
55080
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.