Learn More
Novus Biologicals™ Human vWF-A1 (Research grade Caplacizumab Biosimilar) Antibody, Novus Biologicals™
GREENER_CHOICE
Llama Monoclonal Antibody
Brand:  Novus Biologicals™ MAB10591-SP
 View more versions of this product
Product Code. 18774554
  • ONLINE EXCLUSIVE PRICE
    € 248.00
    € 234.15 / 25µg
    Save € 13.85
    5% Off
Description
vWF-A1 Monoclonal antibody specifically detects vWF-A1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
vWF-A1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
vWF-A1 protein
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Flow Cytometry (Intracellular Staining)
Hu142
Intracellular Staining by Flow Cytometry 0.25 μg/106 cells
Llama
Protein A or G purified from cell culture supernatant
RUO
7450