missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human vWF-A1 (Research grade Caplacizumab Biosimilar) Antibody, Novus Biologicals™

Llama Monoclonal Antibody
€ 248.00 - € 572.00
Specifications
| Antigen | vWF-A1 |
|---|---|
| Clone | Hu142 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 μg/106 cells |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18784384
|
Novus Biologicals™
MAB10591-100 |
100 μg |
€ 572.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18774554
![]() |
Novus Biologicals™
MAB10591-SP |
25 μg |
€ 248.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
vWF-A1 Monoclonal antibody specifically detects vWF-A1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)Specifications
| vWF-A1 | |
| Intracellular Staining by Flow Cytometry 0.25 μg/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| vWF-A1 protein | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Hu142 | |
| Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Llama | |
| Human | |
| 7450 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title