Learn More
Huntingtin Antibody (6F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26212-100ul
 View more versions of this product
Product Code. 30032930
  • € 533.00 / 100µL
Description
Huntingtin Monoclonal antibody specifically detects Huntingtin in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Huntingtin
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
6F2
ELISA, Immunohistochemistry 1:50-1:200
HD protein, huntingtin, huntingtin (Huntington disease), IT15HDHuntington disease protein
A synthesized peptide derived from Human Huntingtin [UniProt P42858]
100 μL
Apoptosis, Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
3064
Store at -20 to -70C. Avoid freeze-thaw cycles.