missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Hyflex™ 11-421 Series Light Duty Nitrile Polyurethane Gloves

Product Code. p-7113232 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 15598595

Brand: Ansell™ 11421100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides high degree of flexibility and durability with optimum dexterity. Ansell Hyflex™ 11-421 Series Light Duty Nitrile Polyurethane Gloves are made from water based mix synthetic compound.

  • Multipurpose light duty gloves
  • Palm fit coating
  • Seamless black nylon liner, gauge 13
  • Fine and stretchable stitches for excellent comfort
  • Silicone-free
  • Not suitable for contact with acidic foodstuffs
  • Offers an indicator button (green) which belong to a range of color code corresponding to a cut level : 1 (green), 3 (orange), 5 (red)

Applications:

  • Delicate manipulation
  • Finishing works
  • General handling
  • Packaging
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2006
Description Gauge: 13
Applications Industrial
Coating Material Polyurethane
Color Gray, Black
Exterior Finish Palm Coated
Latex Free Yes
Material Nitrile
Product Type Mechanical Protection Gloves
Size 10
Liner Material Nylon
Packaging Type 12 pairs in a bag, 12 bags in a carton
Series 11-421
Length (Metric) 220 to 260 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.