missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 47-400 Medium Duty Nitrile Coated Oil Repellent Gloves

Product Code. 10172544 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
144 pairs
This item is not returnable. View return policy

Product Code. 10172544

Brand: Ansell 47400_10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ActivArmr™ gloves combine the comfort of cotton or light leather gloves with the abrasion resistance and oil repellence of nitrile coated gloves.

  • ActivArmr™ gloves feature interlock knit lining, generous sizing, and contoured designs for improved comfort
  • Special nitrile formulation provides a combination of flexibility, mechanical resistance, and oil repellency
  • Natural hand shape is easier to wear for long periods
  • Gloves ensure cleanliness of metal sheets in the steel and motor industries
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Cuff Material Nitrile
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 47-400
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.