Learn More
Ansell HyFlex™ 48-101 Polyurethane Coated Seamless Knit Nylon Gloves
Product Code. 12630922 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 12630922
Brand: Ansell 48101110
Only null left
Add to basket
Edge
Add to basket
Gloves are designed for light applications that require precise handling of small objects in dry or slightly oily environments.
  • Designed for light duty applications that require comfort and abrasion resistance
  • Polyurethane coating enhances grip in dry or light oil applications
  • Seamless construction prevents irritation
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 3
Chlorinated No
Coated Area Palm
Construction Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A1
Exterior Finish Rough
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 11
For Use With (Application) Assembly, Inspection, and Packaging
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Series 48-101
Yarn Gauge 13