Learn More
ICAM-1/CD54 Antibody (3E2B) - Azide and BSA Free, Novus Biologicals™
Hamster Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-22540
 View more versions of this product
Product Code. 18751744
  • € 519.00 / 500µg
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Inhibition Activity, CyTOF-ready, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inhibition Assays, Neutralization
1 mg/mL
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Western Blot 1:10 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Inhibition Activity, CyTOF-ready, Neutralization
ICAM1