Learn More
Fisherbrand™ IEF Conversion Kit
Product Code. 15116634 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Verti-Gel Maxi 2D
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15116634 Verti-Gel Maxi 2D Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15116634
Only null left
Add to basket
Add to basket
Accessory for Verti-Gel Maxi 2D Electrophoresis System.
  • For 18cm IPG strips and tube gels
  • For 2D runs with 180mm IPG strips and gels
Specifications
Description For 2D runs with 180mm IPG strips and gels
For Use With (Equipment) Verti-Gel Maxi 2D
Type IEF Conversion Kit