missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-beta Rabbit anti-Human, Clone: 40, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89612-100UL
This item is not returnable.
View return policy
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for ELISA.
Specifications
| IFN-beta | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 40 | |
| ELISA 1:5000-1:10000 | |
| Fibroblast interferon, IFBIFNB, IFF, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Recombinant Human IFN-beta protein | |
| 100 μg | |
| Apoptosis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3456 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction