missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-beta Rabbit anti-Human, Clone: 40, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 392.00 - € 578.00
Specifications
| Antigen | IFN-beta |
|---|---|
| Clone | 40 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207617
|
Novus Biologicals
NBP2-89612-50UL |
50 μg |
€ 392.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18211956
|
Novus Biologicals
NBP2-89612-100UL |
100 μg |
€ 578.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for ELISA.Specifications
| IFN-beta | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Fibroblast interferon, IFBIFNB, IFF, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Recombinant Human IFN-beta protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 40 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 0.2 um filtered solution in PBS | |
| 3456 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title