missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFNA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121886
This item is not returnable.
View return policy
Description
IFNA4 Polyclonal Antibody for Western Blot
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| IFNA4 | |
| Polyclonal | |
| Unconjugated | |
| IFNA4 | |
| Ifa4; IFNA4; IFN-alpha-4; IFN-alpha4a; INFA4; Interferon; interferon alpha 4; interferon alpha family, gene 4; interferon alpha-4; Interferon alpha-4B; interferon alpha-76; Interferon alpha-M1; interferon, alpha 4; MuIFN-alpha 4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 15967, 3441 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P05014, P07351 | |
| IFNA4 | |
| Recombinant protein Interferon Alpha 4. The antigen corresponds to amino acid range 10-163 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction