missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFNA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 159.00 - € 449.00
Specifications
| Antigen | IFNA4 |
|---|---|
| Concentration | 0.55 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17730624
![]() |
Invitrogen™
PA5119215 |
200 μL |
€ 449.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17743284
|
Invitrogen™
PA5121886 |
20 μL |
€ 159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFNA4 Polyclonal Antibody for Western Blot
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.Spécification
| IFNA4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| IFNA4 | |
| Ifa4; IFNA4; IFN-alpha-4; IFN-alpha4a; INFA4; Interferon; interferon alpha 4; interferon alpha family, gene 4; interferon alpha-4; Interferon alpha-4B; interferon alpha-76; Interferon alpha-M1; interferon, alpha 4; MuIFN-alpha 4 | |
| IFNA4 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.55 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P05014, P07351 | |
| 15967, 3441 | |
| Recombinant protein Interferon Alpha 4. The antigen corresponds to amino acid range 10-163 of the target protein. | |
| Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit