missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig κ Light chain Mouse anti-Human, FITC, Clone: G20-193, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560950
This item is not returnable.
View return policy
Description
The G20-193 monoclonal antibody specifically binds to human immunoglobulin light chain, kappa (κ). It does not react with human immunoglobulin μ light chains or human immunoglobulin heavy chains.
Specifications
| Ig κ Light Chain | |
| Monoclonal | |
| FITC | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| G20-193 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction