missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig κ Light chain Mouse anti-Human, FITC, Clone: G20-193, BD
Mouse Monoclonal Antibody
€ 114.49 - € 212.93
Specifications
| Clone | G20-193 |
|---|---|
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15840629
|
BD Biosciences
560950 |
25 Tests |
€ 115.00 € 114.49 / Each Save € 0.51 0.44% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15868438
|
BD Biosciences
555791 |
100 Tests |
€ 213.00 € 212.93 / Each Save € 0.07 0.03% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The G20-193 monoclonal antibody specifically binds to human immunoglobulin light chain, kappa (κ). It does not react with human immunoglobulin μ light chains or human immunoglobulin heavy chains.
Specifications
| G20-193 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title