missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (V88P3F7*D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50438
This item is not returnable.
View return policy
Description
IgD Monoclonal antibody specifically detects IgD in Fish samples. It is validated for ELISA
Specifications
| IgD | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000 | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Ovalbumin-conjugated synthetic peptide | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3495 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| ELISA | |
| V88P3F7*D2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Fish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction