missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (V88P3F7*D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 407.00
Specifications
| Antigen | IgD |
|---|---|
| Clone | V88P3F7*D2 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600888
|
Novus Biologicals
NBP2-50438-0.025ml |
0.025 mL |
€ 201.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658528
|
Novus Biologicals
NBP2-50438 |
0.1 mL |
€ 407.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgD Monoclonal antibody specifically detects IgD in Fish samples. It is validated for ELISASpecifications
| IgD | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| B Cell Development and Differentiation Markers, Immunology | |
| PBS | |
| 3495 | |
| IgG2b κ | |
| Protein A purified |
| V88P3F7*D2 | |
| ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Fish | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Ovalbumin-conjugated synthetic peptide | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title