missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (V53P3H2*B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50439-0.025ml
This item is not returnable.
View return policy
Description
IgM Monoclonal antibody specifically detects IgM in Fish samples. It is validated for ELISA
Specifications
| IgM | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000 | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Ovalbumin-conjugated synthetic peptide | |
| 0.025 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| ELISA | |
| V53P3H2*B2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Fish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction