missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (V53P3H2*B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.60 - € 406.35
Specifications
| Antigen | IgM |
|---|---|
| Clone | V53P3H2*B2 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693998
|
Novus Biologicals
NBP2-50439-0.025ml |
0.025 mL |
€ 214.00 € 201.60 / 0.03mL Save € 12.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625768
|
Novus Biologicals
NBP2-50439 |
0.1 mL |
€ 430.00 € 406.35 / 0.10mL Save € 23.65 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgM Monoclonal antibody specifically detects IgM in Fish samples. It is validated for ELISASpecifications
| IgM | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| B Cell Development and Differentiation Markers, Immunology | |
| PBS | |
| 3507 | |
| IgG1 κ | |
| Protein A purified |
| V53P3H2*B2 | |
| ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Fish | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Ovalbumin-conjugated synthetic peptide | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title