Learn More
Invitrogen™ IgM Monoclonal Antibody (SA-DA4), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 363-9998-41
This item is not returnable.
View return policy
Description
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications
| Human IgM | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P01871 | |
| IGHM | |
| Affinity chromatography | |
| RUO | |
| 3507 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| SA-DA4 | |
| Brilliant Ultraviolet 395 | |
| IGHM | |
| AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |