missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgM Monoclonal Antibody (SA-DA4), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

€ 216.00 - € 404.00

Specifications

Antigen Human IgM
Clone SA-DA4
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17924342
View Documents
Invitrogen™
363-9998-41
25 Tests
€ 216.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17904162
GREENER_CHOICE
View Documents
Invitrogen™
363-9998-42
100 Tests
€ 404.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications

Specifications

Human IgM
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH
IGHM
Primary
4°C, store in dark, DO NOT FREEZE!
SA-DA4
Flow Cytometry
Brilliant Ultraviolet 395
Mouse
Human
P01871
3507
IgG1 κ
Affinity chromatography
IGHM
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.