Learn More
Invitrogen™ IgM Monoclonal Antibody (SA-DA4), Brilliant Violet™ 786, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ 417-9998-41
 View more versions of this product
Product Code. 17951991
  • € 193.00 / Pack of 25
Description
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgM
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P01871
IGHM
Affinity chromatography
RUO
3507
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
SA-DA4
Brilliant Violet 786
IGHM
AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH
Mouse
25 Tests