missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgM Monoclonal Antibody (SA-DA4), Brilliant Violet™ 786, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
€ 188.00 - € 385.00
Specifications
| Antigen | Human IgM |
|---|---|
| Clone | SA-DA4 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17951991
|
Invitrogen™
417-9998-41 |
25 Tests |
€ 188.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17992191
![]() |
Invitrogen™
417-9998-42 |
100 Tests |
€ 385.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.Specifications
| Human IgM | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH | |
| IGHM | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| SA-DA4 | |
| Flow Cytometry | |
| Brilliant Violet 786 | |
| Mouse | |
| Human | |
| P01871 | |
| 3507 | |
| IgG1 κ | |
| Affinity chromatography | |
| IGHM |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title