missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin 6 Antibody (RM0157-3L27) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12423
This item is not returnable.
View return policy
Description
Cathepsin 6 Monoclonal specifically detects Cathepsin 6 in Mouse samples. It is validated for Western Blot.
Specifications
| Cathepsin 6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| NP_067420 | |
| Cts6 | |
| Mouse recombinant Cathepsin 6 | |
| 0.1 mg | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot | |
| RM0157-3L27 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| 1600022N02Rik | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 58518 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction