missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IKK 16

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective inhibitor of IκB kinase (IKK) (IC50 values are 40, 70 and 200 nM for IKKβ, IKK complex and IKKα respectively). Inhibits TNF-α-stimulated IκB degradation and expression of adhesion molecules E-selectin, ICAM and VCAM (IC50 values are 1.0, 0.5, 0.3 and 0.3 μM respectively). Inhibits LPS-induced TNF-α release in vivo and neutrophil extravasion in thioglycollate-induced peritonitis.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 520.09 |
| Purity | >98% |
| Molecular Weight (g/mol) | 520.092 |
| Chemical Name or Material | IKK 16 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction