Learn More
IL-12 p70 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen 500-P154HGBT-1MG
Description
AA Seq: p35 Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS. p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS Preparation: Produced from sera of goats immunized with highly pure Recombinant Human IL-12. Anti-Human IL-12-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-12 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-12 (500-P154HG) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human IL-12. Western Blot: To detect Human IL-12 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/ml. When used in conjunction with compatible development reagents the detection limit for Recombinant Human IL-12 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
Specifications
| IL-12 p70 | |
| Polyclonal | |
| PBS with no preservative | |
| P29459, P29460 | |
| IL12A, Il12b | |
| HEK293 cells-derived Recombinant Human IL-12 p70 | |
| 2 x 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
| ELISA, Western Blot | |
| Biotin | |
| Il12b | |
| 20C2, CLMF, CLMF p35, CLMF p40, CLMF2, cytokine, cytotoxic lymphocyte maturation factor 1, p35, cytotoxic lymphocyte maturation factor 35 kDa subunit, Cytotoxic lymphocyte maturation factor 40 kDa subunit, il 12, IL 12p70, Il12, IL-12, Il-12 p35, IL-12 p75, IL-12 subunit p35, IL-12 subunit p40, IL-12, subunit p35, IL12, subunit p40, Il-12, cytokine, IL12A, IL-12A, IL12A/B, IL12AB, IL12B, Il-12b, IL12P35, IL-12p35, Il12p40, Il-12p40, IL35 subunit, ILN, IMD28, IMD29, Interleukin, interleukin 12 35 kDa subunit, interleukin 12 35kDa subunit, interleukin 12 40 kDa subunit, interleukin 12 40kDa subunit, interleukin 12 p35 subunit, interleukin 12 p40 subunit precursor, interleukin 12 polypeptide 35, interleukin 12, p35, interleukin 12, p40, interleukin 12A, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35), interleukin 12B, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40), interleukin 12p35 subunit precursor, interleukin-12 alpha chain, interleukin-12 alpha p40 subunit, interleukin-12 beta chain, interleukin-12 beta p35 subunit, interleukin-12 p35 subunit, interleukin-12 p35 subunit precursor, interleukin-12 p40, interleukin-12 p40 subunit, interleukin-12 p40 subunit precursor, Interleukin-12 p70, interleukin-12 subunit alpha, Interleukin-12 subunit beta, interleukin-23 p40 subunit, Ll12a, MGC151228, MGC151232, natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35, natural killer cell stimulatory factor, 40 kD subunit, NF cell stimulatory factor chain 1, NFSK, NK cell stimulatory factor chain 1, NK cell stimulatory factor chain 2, NKSF, NKSF1, NKSF2, P35, p40 | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| 3592, 3593 | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.