missing translation for 'onlineSavingsMsg'
Learn More

IL-12 p70 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

€ 227.00 - € 3435.00

Specifications

Antigen IL-12 p70
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Form Lyophilized
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17886397
View Documents
Invitrogen
500-P154HGBT-1MG
2 x 500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17846547
View Documents
PeproTech
500-P154HGBT-25UG
25 μg
€ 227.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17856567
View Documents
PeproTech
500-P154HGBT-50UG
50 μg
€ 284.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30054526
View Documents
PeproTech
500-P154HGBT-500UG
500 μg
€ 1884.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Seq: p35 Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS. p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS Preparation: Produced from sera of goats immunized with highly pure Recombinant Human IL-12. Anti-Human IL-12-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-12 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-12 (500-P154HG) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human IL-12. Western Blot: To detect Human IL-12 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/ml. When used in conjunction with compatible development reagents the detection limit for Recombinant Human IL-12 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
Specifications

Specifications

IL-12 p70
Polyclonal
Lyophilized
RUO
PBS with no preservative
20C2, CLMF, CLMF p35, CLMF p40, CLMF2, cytokine, cytotoxic lymphocyte maturation factor 1, p35, cytotoxic lymphocyte maturation factor 35 kDa subunit, Cytotoxic lymphocyte maturation factor 40 kDa subunit, il 12, IL 12p70, Il12, IL-12, Il-12 p35, IL-12 p75, IL-12 subunit p35, IL-12 subunit p40, IL-12, subunit p35, IL12, subunit p40, Il-12, cytokine, IL12A, IL-12A, IL12A/B, IL12AB, IL12B, Il-12b, IL12P35, IL-12p35, Il12p40, Il-12p40, IL35 subunit, ILN, IMD28, IMD29, Interleukin, interleukin 12 35 kDa subunit, interleukin 12 35kDa subunit, interleukin 12 40 kDa subunit, interleukin 12 40kDa subunit, interleukin 12 p35 subunit, interleukin 12 p40 subunit precursor, interleukin 12 polypeptide 35, interleukin 12, p35, interleukin 12, p40, interleukin 12A, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35), interleukin 12B, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40), interleukin 12p35 subunit precursor, interleukin-12 alpha chain, interleukin-12 alpha p40 subunit, interleukin-12 beta chain, interleukin-12 beta p35 subunit, interleukin-12 p35 subunit, interleukin-12 p35 subunit precursor, interleukin-12 p40, interleukin-12 p40 subunit, interleukin-12 p40 subunit precursor, Interleukin-12 p70, interleukin-12 subunit alpha, Interleukin-12 subunit beta, interleukin-23 p40 subunit, Ll12a, MGC151228, MGC151232, natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35, natural killer cell stimulatory factor, 40 kD subunit, NF cell stimulatory factor chain 1, NFSK, NK cell stimulatory factor chain 1, NK cell stimulatory factor chain 2, NKSF, NKSF1, NKSF2, P35, p40
IL12A, Il12b
Primary
-20°C
ELISA, Western Blot
Biotin
Goat
Human
P29459, P29460
3592, 3593
HEK293 cells-derived Recombinant Human IL-12 p70
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.