missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17/IL-17A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11319-0.025mg
This item is not returnable.
View return policy
Description
IL-17/IL-17A Polyclonal antibody specifically detects IL-17/IL-17A in Guinea Pig samples. It is validated for ELISA
Specifications
| IL-17/IL-17A | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Guinea Pig | |
| Purified |
| ELISA | |
| Unconjugated | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Recombinant guinea pig IL-17A | |
| 0.025 mg | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| 3605 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction