missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17/IL-17A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 287.00 - € 840.00
Specifications
| Antigen | IL-17/IL-17A |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630109
|
Novus Biologicals
NBP3-11319 |
0.1 mg |
€ 840.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667046
|
Novus Biologicals
NBP3-11319-0.025mg |
0.025 mg |
€ 287.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17/IL-17A Polyclonal antibody specifically detects IL-17/IL-17A in Guinea Pig samples. It is validated for ELISASpecifications
| IL-17/IL-17A | |
| Polyclonal | |
| Purified | |
| RUO | |
| Guinea Pig | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Recombinant guinea pig IL-17A | |
| Primary | |
| Store at 4°C. Do not freeze. |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| PBS | |
| 3605 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title