missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Rat anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: MQ1-17H12, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566406
This item is not returnable.
View return policy
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
Specifications
| IL-2 | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 25 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
| Flow Cytometry | |
| MQ1-17H12 | |
| Rat | |
| RUO | |
| Human, Baboon, Cynomolgus, Rhesus Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction