missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Rat anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: MQ1-17H12, BD Horizon™
Rat Monoclonal Antibody
€ 225.05 - € 406.00
Specifications
| Antigen | IL-2 |
|---|---|
| Clone | MQ1-17H12 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15895019
|
BD Biosciences
566406 |
25 Tests |
€ 232.00 € 225.05 / Each Save € 6.95 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15885019
|
BD Biosciences
566405 |
100 Tests |
€ 406.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Specifications
| IL-2 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Rat | |
| Human, Baboon, Cynomolgus, Rhesus Macaque | |
| Primary |
| MQ1-17H12 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title