missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ILT8/CD85b/LILRA6 Antibody (1B5B11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27159-0.025ml
This item is not returnable.
View return policy
Description
ILT8/CD85b/LILRA6 Monoclonal antibody specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications
| ILT8/CD85b/LILRA6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| CD85b;ILT5;ILT8;ILT-8;LILRB3;LILRB6 | |
| Purified recombinant fragment of human ILT8/CD85b/LILRA6 (AA: 52-195) expressed in E. Coli. | |
| 0.025 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Western Blot | |
| 1B5B11 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 79168 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction