missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ILT8/CD85b/LILRA6 Antibody (1B5B11), Novus Biologicals™
Mouse Monoclonal Antibody
€ 298.00 - € 511.00
Specifications
| Antigen | ILT8/CD85b/LILRA6 |
|---|---|
| Clone | 1B5B11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30026009
|
Novus Biologicals™
NBP3-27159-0.1ml |
0.1 mL |
€ 511.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30031345
|
Novus Biologicals™
NBP3-27159-0.025ml |
0.025 mL |
€ 298.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILT8/CD85b/LILRA6 Monoclonal antibody specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| ILT8/CD85b/LILRA6 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD85b;ILT5;ILT8;ILT-8;LILRB3;LILRB6 | |
| Purified recombinant fragment of human ILT8/CD85b/LILRA6 (AA: 52-195) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1B5B11 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 79168 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title