missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAS1L Antibody (15B8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26148-50ul
This item is not returnable.
View return policy
Description
MAS1L Monoclonal antibody specifically detects MAS1L in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Specifications
| MAS1L | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| 15B8 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| dJ994E9.2, MAS1 oncogene-like, MAS1-like, MAS-L, MAS-R, mas-related G protein-coupled MRG, mas-related G-protein coupled receptor MRG, MRGMGC119987 | |
| A synthesized peptide derived from Human MAS1L [UniProt P35410] | |
| 50 μL | |
| GPCR | |
| 116511 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction