missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP2/IGF2BP2 Antibody (SR2110), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22430-100ul
This item is not returnable.
View return policy
Description
IMP2/IGF2BP2 Monoclonal antibody specifically detects IMP2/IGF2BP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| IMP2/IGF2BP2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2110 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2 | |
| A synthesized peptide derived from human IMP2/IGF2BP2 (Uniprot #: Q9Y6M1) | |
| 100 μg | |
| Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 10644 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction