missing translation for 'onlineSavingsMsg'
Learn More

IMP2/IGF2BP2 Antibody (SR2110), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 338.00 - € 449.00

Specifications

Antigen IMP2/IGF2BP2
Clone SR2110
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18659613
View Documents Promotion Details
Novus Biologicals
NBP3-22430-100ul
100 μg
€ 449.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18640755
View Documents
Novus Biologicals
NBP3-22430-50ul
50 μg
€ 338.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IMP2/IGF2BP2 Monoclonal antibody specifically detects IMP2/IGF2BP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

IMP2/IGF2BP2
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2
A synthesized peptide derived from human IMP2/IGF2BP2 (Uniprot #: Q9Y6M1)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2110
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
10644
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.