Learn More
INCB3344, MedChemExpress
Product Code. 16477846
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16477846
Brand: Medchem Express HY50674
MedChemExpress INCB3344 is a potent, selective and orally bioavailable CCR2 antagonist with IC50 values of 5.1 nM (hCCR2) and 9.5 nM (mCCR2) in binding antagonism and 3.8 nM (hCCR2) and 7.8 nM (mCCR2) in antagonism of chemotaxis activity.
INCB3344 is a potent, selective and orally bioavailable CCR2 antagonist with IC50 values of 5.1 nM (hCCR2) and 9.5 nM (mCCR2) in binding antagonism and 3.8 nM (hCCR2) and 7.8 nM (mCCR2) in antagonism of chemotaxis activity.
Specifications
Chemical Name or Material INCB3344
CAS 1262238-11-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H34F3N3O6
Quantity 100 mg
Solubility Information DMSO : 220 mg/mL (380.89 mM; Need ultrasonic)
SMILES CCO[C@H]1CN([C@]2([H])CC[C@@](CC2)(C3=CC4=C(OCO4)C=C3)O)C[C@@H]1NC(CNC(C5=CC(C(F)(F)F)=CC=C5)=O)=O
Molecular Weight (g/mol) 577.59
Formula Weight 577.59
Percent Purity 96.59%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only