Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2
Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
Specifications
Specifications
| Classification | F2 |
| Material | Stainless Steel |
| Weight (Metric) | 2 g |
| Mass | 2 g |
| Shape | Cylindrical |