missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 M2 Antibody (14C2) - (A/WSN/1933), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2073-0.025mg
This item is not returnable.
View return policy
Description
Influenza A H1N1 M2 Monoclonal antibody specifically detects Influenza A H1N1 M2 in Influenza A Virus H1N1 samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Block/Neutralize.
Specifications
| Influenza A H1N1 M2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 10 μg/mL, Neutralization | |
| M2 protein from A/WSN/33-infected CV1 cell lysate. | |
| 0.025 mg | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| 14C2 | |
| Unconjugated | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Influenza A virus H1N1 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction