missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 M2 Antibody (14C2) - (A/WSN/1933), Novus Biologicals™
Mouse Monoclonal Antibody
€ 213.00
Specifications
| Antigen | Influenza A H1N1 M2 |
|---|---|
| Clone | 14C2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 10 μg/mL, Neutralization |
| Applications | Western Blot |
Description
Influenza A H1N1 M2 Monoclonal antibody specifically detects Influenza A H1N1 M2 in Influenza A Virus H1N1 samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Block/Neutralize.Specifications
| Influenza A H1N1 M2 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Influenza A virus H1N1 | |
| IgG1 κ | |
| Protein A purified |
| 14C2 | |
| Western Blot 1 μg/mL, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 10 μg/mL, Neutralization | |
| Monoclonal | |
| Purified | |
| RUO | |
| M2 protein from A/WSN/33-infected CV1 cell lysate. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title