missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H3N2 Hemagglutinin Antibody - (A/Perth/16/2009), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12731-50ul
This item is not returnable.
View return policy
Description
Influenza A H3N2 Hemagglutinin Polyclonal antibody specifically detects Influenza A H3N2 Hemagglutinin in Influenza A virus H3N2 samples. It is validated for Western Blot
Specifications
| Influenza A H3N2 Hemagglutinin | |
| Polyclonal | |
| Western Blot 1:1000-1:5000 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Influenza A H3N2 Hemagglutinin (Accession#: ADI52838.1; Met1-Arg345). | |
| 50 μg | |
| Virology Bacteria and Parasites | |
| Influenza A virus H3N2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction