missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H3N2 Hemagglutinin Antibody - (A/Perth/16/2009), Novus Biologicals™
Rabbit Polyclonal Antibody
€ 299.00 - € 507.00
Specifications
| Antigen | Influenza A H3N2 Hemagglutinin |
|---|---|
| Dilution | Western Blot 1:1000-1:5000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688717
|
Novus Biologicals
NBP3-12731-100ul |
100 μg |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699446
|
Novus Biologicals
NBP3-12731-50ul |
50 μg |
€ 299.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza A H3N2 Hemagglutinin Polyclonal antibody specifically detects Influenza A H3N2 Hemagglutinin in Influenza A virus H3N2 samples. It is validated for Western BlotSpecifications
| Influenza A H3N2 Hemagglutinin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:1000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Influenza A virus H3N2 | |
| Produced in rabbits immunized with purified, recombinant Influenza A H3N2 Hemagglutinin (Accession#: ADI52838.1; Met1-Arg345). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title