Learn More
Influenza A H5N1 Hemagglutinin Antibody (1E6A7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-75493MFV500
 View more versions of this product
Product Code. 18477057
  • € 588.00 / 0.10mL
Description
Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza A H5N1 Hemagglutinin
Monoclonal
mFluor Violet 500 SE
Mouse
Protein A purified
RUO
Virus
Purified
Western Blot
1E6A7
50mM Sodium Borate
A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein.
0.1 mL
Primary
Store at 4°C in the dark.