missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Antibody (1E6A7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
€ 588.00
Specifications
| Antigen | Influenza A H5N1 Hemagglutinin |
|---|---|
| Clone | 1E6A7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 500 SE |
Description
Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
| Influenza A H5N1 Hemagglutinin | |
| Western Blot | |
| mFluor Violet 500 SE | |
| Mouse | |
| Virus | |
| A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein. | |
| Primary | |
| Store at 4°C in the dark. |
| 1E6A7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title