Learn More
Influenza A H5N1 Hemagglutinin Antibody (7B9B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-75495-0.025mg
This item is not returnable.
View return policy
Description
Influenza A Hemagglutinin Monoclonal specifically detects Influenza A Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Influenza A Hemagglutinin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| HA epitope tag, HA1, HA2, Hemagglutinin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Virus |
| Western Blot | |
| 7B9B2 | |
| Unconjugated | |
| AAT76166 | |
| HA | |
| A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein. | |
| 0.025 mg | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |