Learn More
Influenza A H5N1 Hemagglutinin Antibody (7B9B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-75495-0.025mg
 View more versions of this product
Product Code. 18595534
  • € 209.00 / 25µg
Description
Influenza A Hemagglutinin Monoclonal specifically detects Influenza A Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza A Hemagglutinin
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
HA epitope tag, HA1, HA2, Hemagglutinin
Mouse
Affinity Purified
RUO
Virus
Western Blot
7B9B2
Unconjugated
AAT76166
HA
A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein.
0.025 mg
Primary
Store at -20°C. Avoid freeze/thaw cycles.