missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Antibody (7B9B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 209.00
Specifications
| Antigen | Influenza A Hemagglutinin |
|---|---|
| Clone | 7B9B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot |
Description
Influenza A Hemagglutinin Monoclonal specifically detects Influenza A Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.Specifications
| Influenza A Hemagglutinin | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| AAT76166 | |
| HA | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 7B9B2 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Virus | |
| HA epitope tag, HA1, HA2, Hemagglutinin | |
| A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein. | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title