missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A nucleoprotein Antibody (7B4G10G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23452-100ul
This item is not returnable.
View return policy
Description
Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza A nucleoprotein | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 7B4G10G8 | |
| Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 | |
| Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein | |
| This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction