missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A nucleoprotein Antibody (7B4G10G8), Novus Biologicals™
Mouse Monoclonal Antibody
€ 195.00 - € 507.00
Specifications
| Antigen | Influenza A nucleoprotein |
|---|---|
| Clone | 7B4G10G8 |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664314
|
Novus Biologicals
NBP3-23452-100ul |
100 μg |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636414
|
Novus Biologicals
NBP3-23452-20ul |
20 μg |
€ 195.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Virus samples. It is validated for Western Blot, ELISASpecifications
| Influenza A nucleoprotein | |
| Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 7B4G10G8 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title